Merck
  • Effect of chemical modifications on peptide fragmentation behavior upon electron transfer induced dissociation.
Effect of chemical modifications on peptide fragmentation behavior upon electron transfer induced dissociation.
Analytical chemistry (2009-08-20)
Marco L Hennrich, Paul J Boersema, Henk van den Toorn, Nikolai Mischerikow, Albert J R Heck, Shabaz Mohammed
ABSTRACT
In proteomics, proteolytic peptides are often chemically modified to improve MS analysis, peptide identification, and/or to enable protein/peptide quantification. It is known that such chemical modifications can alter peptide fragmentation in collision induced dissociation MS/MS. Here, we investigated the fragmentation behavior of such chemically modified peptides in MS/MS using the relatively new activation method electron transfer dissociation (ETD). We generated proteolytic peptides using the proteases Lys-N and trypsin and compared the fragmentation behavior of the unlabeled peptides with that of their chemically modified cognates. We investigated the effect of several commonly used modification reactions, namely, guanidination, dimethylation, imidazolinylation, and nicotinylation (ICPL). Of these guanidination and imidazolinylation specifically target the epsilon-amino groups of lysine residues in the peptides, whereas dimethylation and nicotinylation modify both N-termini and epsilon-amino groups of lysine residues. Dimethylation, guanidination, and particularly imidazolinylation of doubly charged Lys-N peptides resulted in a significant increase in peptide sequence coverage, resulting in more reliable peptide identification using ETD. This may be rationalized by the increased basicity and resulting positive charge at the N-termini of these peptides. Nicotinylation of the peptides, on the other hand, severely suppressed backbone fragmentation, hampering the use of this label in ETD based analysis. Doubly charged C-terminal lysine containing tryptic peptides also resulted in an enhanced observation of a single type of fragment ion series when guanidinated or imidazolinylated. These labels would thus facilitate the use of de novo sequencing strategies based on ETD for both arginine and lysine containing tryptic peptides. Since isotopic analogues of the labeling reagents applied in this work are commercially available, one can combine quantitation with improved ETD based peptide sequencing for both Lys-N and trypsin digested samples.
Sigma-Aldrich
Triethylammonium bicarbonate buffer, volatile buffer, ~1.0 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Triethylammonium bicarbonate buffer, 1.0 M, pH 8.5±0.1
Pricing and availability is not currently available.