Merck
  • Metal-ion-induced folding and dimerization of a glycoluril decamer in water.
Metal-ion-induced folding and dimerization of a glycoluril decamer in water.
Organic letters (2009-08-07)
Wei-Hao Huang, Peter Y Zavalij, Lyle Isaacs
ABSTRACT
In a process reminiscent of RNA folding, acyclic glycoluril decamer (+/-)-1 undergoes double helical assembly (red/green strands) in water triggered by the neutralization of regions of high electrostatic surface potential by metal cations (Li(+), Na(+), K(+), Ca(2+)).
Sigma-Aldrich
Glycoluril, 97%
Pricing and availability is not currently available.