- Directed carbon nanotube assembly using a pyrene-functionalized polymer.
Directed carbon nanotube assembly using a pyrene-functionalized polymer.
Chemical communications (Cambridge, England) (2009-08-05)
Eric Schopf, Rebecca Broyer, Lei Tao, Yong Chen, Heather D Maynard
PMID19652791
ABSTRACT
A pyrene-functionalized polymer was patterned via electron beam lithography onto a silicon wafer and shown to selectively bind with carbon nanotubes.