Merck
  • Optimization of a method for determination of phenolic acids in exotic fruits by capillary electrophoresis.
Optimization of a method for determination of phenolic acids in exotic fruits by capillary electrophoresis.
Journal of pharmaceutical and biomedical analysis (2009-06-24)
Tatiana S Fukuji, Fernando G Tonin, Marina F M Tavares
ABSTRACT
In this work, the separation of nine phenolic acids (benzoic, caffeic, chlorogenic, p-coumaric, ferulic, gallic, protocatechuic, syringic, and vanillic acid) was approached by a 3(2) factorial design in electrolytes consisting of sodium tetraborate buffer (STB) in the concentration range of 10-50 mmol L(-1) and methanol in the volume percentage of 5-20%. Derringer's desirability functions combined globally were tested as response functions. An optimal electrolyte composed by 50 mmol L(-1) tetraborate buffer at pH 9.2, and 7.5% (v/v) methanol allowed baseline resolution of all phenolic acids under investigation in less than 15 min. In order to promote sample clean up, to preconcentrate the phenolic fraction and to release esterified phenolic acids from the fruit matrix, elaborate liquid-liquid extraction procedures followed by alkaline hydrolysis were performed. The proposed methodology was fully validated (linearity from 10.0 to 100 microg mL(-1), R(2)>0.999; LOD and LOQ from 1.32 to 3.80 microg mL(-1) and from 4.01 to 11.5 microg mL(-1), respectively; intra-day precision better than 2.8% CV for migration time and 5.4% CV for peak area; inter-day precision better than 4.8% CV for migration time and 4.8-11% CV for peak area; recoveries from 81% to 115%) and applied successfully to the evaluation of phenolic contents of abiu-roxo (Chrysophyllum caimito), wild mulberry growing in Brazil (Morus nigra L.) and tree tomato (Cyphomandra betacea). Values in the range of 1.50-47.3 microg g(-1) were found, with smaller amounts occurring as free phenolic acids.
Sigma-Aldrich
Borax Anhydrous, puriss. p.a., anhydrous, ≥98.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Borax Anhydrous, BioUltra, anhydrous, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium tetraborate, 99.998% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium tetraborate, 99%
Pricing and availability is not currently available.