Merck
  • Solubilization of n-alkylbenzene and n-perfluoroalkylbenzene in hydrogenated and fluorinated surfactants micelles.
Solubilization of n-alkylbenzene and n-perfluoroalkylbenzene in hydrogenated and fluorinated surfactants micelles.
Journal of colloid and interface science (2009-02-27)
Keisuke Matsuoka, Emi Takeuchi, Megumi Takahashi, Satomi Kitsugi, Chikako Honda, Kazutoyo Endo
ABSTRACT
The maximum solubilities of homologous series of n-alkylbenzene and n-perfluoroalkylbenzene in aqueous solutions of surfactants n-tetradecyltrimethylammonium chloride (TTAC) and N-(1,1-dihydroperfluorodecyl)-N,N,N-trimethylammonium chloride (C10F-TAC) were measured as a function of the surfactant concentration at 298.2 K. There are four solubilization systems in viewpoints of hydrocarbons and fluorocarbons. In general, in all systems, the maximum solubility decreased with an increase in the alkyl carbon length of the solubilizates. The homogeneous combinations of solubilizates and solubilizers have higher affinity than heterogeneous combinations. The affinity between a hydrocarbon and a fluorocarbon can be clarified by determining the Gibbs free energy (DeltaG(0)) on solubilization by thermodynamic analysis. The largest negative value of DeltaG(0) was obtained for the homogeneous system of n-perfluoroalkylbenzene and C10F-TAC, whereas the smallest value was obtained for the heterogeneous system of n-perfluoroalkylbenzene and TTAC. The contributions of methylene and perfluoromethylene to Gibbs energy, namely, DeltaG(CH(2)0)) and DeltaG(CF(2)(0)), were found to be -2.6 and -2.9 kJ mol(-1), respectively, for the TTAC solution, whereas the respective values for the C10F-TAC solution were -2.0 and -3.3 kJ mol(-1).
Product Description
Supelco
Myristyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0% (AT)
Sigma-Aldrich
Myristyltrimethylammonium bromide, 98% (AT)
Sigma-Aldrich
Trimethyl-tetradecylammonium chloride, ≥98.0% (AT)
Sigma-Aldrich
Myristyltrimethylammonium bromide, ≥99%