Merck
  • Structure of the buried metal-molecule interface in organic thin film devices.
Structure of the buried metal-molecule interface in organic thin film devices.
Nano letters (2009-02-10)
Christian R Hansen, Thomas J Sørensen, Magni Glyvradal, Jacob Larsen, Sara H Eisenhardt, Thomas Bjørnholm, Martin M Nielsen, Robert Feidenhans'l, Bo W Laursen
ABSTRACT
By use of specular X-ray reflectivity (XR) the structure of a metal-covered organic thin film device is measured with angstrom resolution. The model system is a Langmuir-Blodgett (LB) film, sandwiched between a silicon substrate and a top electrode consisting of 25 A titanium and 100 A aluminum. By comparison of XR data for the five-layer Pb2+ arachidate LB film before and after vapor deposition of the Ti/Al top electrode, a detailed account of the structural damage to the organic film at the buried metal-molecule interface is obtained. We find that the organized structure of the two topmost LB layers (approximately 5 nm) is completely destroyed due to the metal deposition.
Sigma-Aldrich
Arachidic acid, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Arachidic acid, synthetic, ≥99.0% (GC)
Pricing and availability is not currently available.