Merck
  • Effects of environmental novelty on fear-related behavior and stress responses of rats to emotionally relevant odors.
Effects of environmental novelty on fear-related behavior and stress responses of rats to emotionally relevant odors.
Behavioural brain research (2008-12-24)
Y Nikaido, T Nakashima
ABSTRACT
Although various emotional behaviors and activation of the hypothalamic-pituitary-adrenal (HPA) axis of rats are induced by the exposure of 2,5-dihydro-2,4,5-trimethylthiazoline: TMT, a component of fox odor, these odor-induced responses are influenced by the external environment. Our previous study demonstrated that exposure to green odor, a mixture of cis-3-hexenol and trans-2-hexenal, attenuated stress-induced elevation of the plasma ACTH level in rats. The present study investigated the effect of TMT or green odor on emotional behavior and the HPA axis stress response with or without the influence of environmental novelty. We exposed rats to TMT or green odor in "familiar" or "unfamiliar" environments and compared the various responses, including fear-related behaviors, non-defensive behaviors and plasma ACTH concentrations. TMT induced enhanced freezing behavior, reduced exploration behavior and elevations in plasma ACTH concentrations in two environmental conditions. Comparing TMT-induced responses in an unfamiliar environment with the familiar environment showed that environmental novelty enhanced TMT-induced fear-related behaviors and elevations of plasma ACTH concentrations. These results revealed that TMT causes fear and stress responses in both familiar and unfamiliar environments, although the novelty of an unfamiliar environment enhances these TMT-induced responses. On the other hand, green odor did not induce any responses in either environment. These findings indicate that odor-induced responses are influenced by the surrounding environment.
Sigma-Aldrich
trans-3-Hexen-1-ol, ≥95%, stabilized
Sigma-Aldrich
trans-3-Hexen-1-ol, 97%
Sigma-Aldrich
cis-3-Hexen-1-ol, 98%
Sigma-Aldrich
cis-3-Hexen-1-ol, natural, >98%, FCC, FG
Supelco
cis-3-Hexen-1-ol, analytical standard