Merck
  • Awakening a dormant catalyst: salicylaldimine systems for ethene/tert-butylstyrene copolymerization.
Awakening a dormant catalyst: salicylaldimine systems for ethene/tert-butylstyrene copolymerization.
Dalton transactions (Cambridge, England : 2003) (2008-12-04)
Giles W Theaker, Colin Morton, Peter Scott
ABSTRACT
A group of readily available zirconium catalysts incapable of ethene-co-styrene polymerization are remarkably active and selective for the production of the new polymer ethene-co-tert-butylstyrene via a single site mechanism.
Sigma-Aldrich
Zirconium(IV) chloride, ≥99.5% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Zirconium(IV) chloride, ≥99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Zirconium(IV) chloride, anhydrous, powder, 99.99% trace metals basis
Pricing and availability is not currently available.