Merck
  • [Major gastrointestinal syndromes (2): dyspepsia or syndrome of the upper digestive system related with food (2)].
[Major gastrointestinal syndromes (2): dyspepsia or syndrome of the upper digestive system related with food (2)].
Revista de gastroenterologia del Peru : organo oficial de la Sociedad de Gastroenterologia del Peru (2008-10-30)
Raúl León-Barúa, Roberto Berendson-Seminario
ABSTRACT
We expose, this time, our ideas and findings on the etiopathogenesis, diagnosis and treatment of the dyspeptic syndrome.