Merck
  • Studies on optical absorption and photoluminescence of thioglycerol-stabilized CdS quantum dots.
Studies on optical absorption and photoluminescence of thioglycerol-stabilized CdS quantum dots.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2008-06-11)
C Unni, Daizy Philip, K G Gopchandran
ABSTRACT
Nanoparticles of CdS were prepared at 303 K by aqueous precipitation method using CdSO4 and (NH4)2S in presence of the stabilizing agent thioglycerol. Adjustment of the thioglycerol (T) to ammonium sulphide (A) ratio (T:A) from 1:25 to 1:3.3 was done during synthesis and nanoparticles of different size were obtained. The prepared colloids were characterized by UV-vis and photoluminescence (PL) spectroscopic studies. Prominent first and second excitonic transitions are observed in the UV-vis spectrum of the colloid prepared with a T:A ratio of 1:3.3. Particle size analysis was done using XRD, high resolution TEM and dynamic light scattering and found to be approximately 3 nm. UV-vis and PL spectral features also agree with this particle size in colloid with T:A of 1:3.3. The band gap of CdS quantum dots has increased from the bulk value 2.4-2.9 eV. PL spectra show quantum size effect and the peak is shifted from 628 to 556 nm when the ratio of T:A was changed from 1:25 to 1:3.3. Doping of CdS with Zn2+ and Cu2+ is found to enhance the PL intensity. PL band shows blue-shift and red-shift on doping with Zn2+ and Cu2+, respectively. UV and PL spectral features of the CdS/Au hybrid nanoparticles obtained by a physical mixing of CdS and Au nanoclusters in various volume ratios is also discussed. Au red-shifts and rapidly quenches the PL of CdS. An additional low energy band approximately 650 nm is observed in the UV visible spectrum of the hybrid nanoparticles.
Sigma-Aldrich
1-Thioglycerol, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
1-Thioglycerol, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
1-Thioglycerol, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Thioglycerol, BioXtra, ≥97%
Pricing and availability is not currently available.