Merck
Catalytic asymmetric epoxidation of cyclic enones.
Journal of the American Chemical Society (2008-04-22)
Xingwang Wang, Corinna M Reisinger, Benjamin List
ABSTRACT
A highly enantioselective epoxidation of cyclic enones with hydrogen peroxide has been developed that is catalyzed by chiral primary amine salts.
Sigma-Aldrich
2-Cyclohexen-1-one, ≥95%