Merck
  • CuCl-mediated tandem CO insertion and annulation of 1,4-dilithio-1,3-dienes: formation of multiply substituted cyclopentadienones and/or their head-to-head dimers.
CuCl-mediated tandem CO insertion and annulation of 1,4-dilithio-1,3-dienes: formation of multiply substituted cyclopentadienones and/or their head-to-head dimers.
Chemical communications (Cambridge, England) (2008-03-21)
Qian Luo, Chao Wang, Wen-Xiong Zhang, Zhenfeng Xi
ABSTRACT
Mediated by CuCl, 1,4-dilithio-1,3-dienes reacted with carbon monoxide (CO) to generate multi-substituted cyclopentadienones and/or their head-to-head dimers, via tandem CO insertion and intra/intermolecular cycloaddition of organocopper compounds.
Sigma-Aldrich
Copper(I) chloride, ReagentPlus®, purified, ≥99%
Sigma-Aldrich
Copper(I) chloride, reagent grade, 97%
Sigma-Aldrich
Copper(I) chloride, AnhydroBeads, ≥99.99% trace metals basis
Sigma-Aldrich
Copper(I) chloride, ≥99.995% trace metals basis