Merck
  • Structural and spectroscopic studies of some copper(I) halide tert-butyl isocyanide adducts.
Structural and spectroscopic studies of some copper(I) halide tert-butyl isocyanide adducts.
Dalton transactions (Cambridge, England : 2003) (2008-03-21)
Graham A Bowmaker, John V Hanna, F Ekkehardt Hahn, Andrew S Lipton, Carolyn E Oldham, Brian W Skelton, Mark E Smith, Allan H White
ABSTRACT
Single-crystal structural characterizations confirm the existence of the unusual 1 : 4 copper(I) halide : unidentate ligand adducts [Cu(CNt-Bu)4]X for X = Cl, Br (two forms), I (the chloride and one form of the bromide being solvated) with crystal packing dominated by stacks of interleaving cations. Cu-C range between 1.941(2) and 1.972(4) A. The structure of the 1 : 2 chloride complex is also recorded, being [ClCu(CNt-Bu)2], with the copper(I) atom environment trigonal planar, while CuCN : (CNt-Bu) (1 : 1) is a single-stranded polymer which spirals about a crystallographic 3-axis (CN scrambled), the ligands being pendant from the ...CuCNCuCN... string. The (5Cu static broadline NMR spectra of [Cu(CNt-Bu)4]I and [Cu(CNt-Bu)4]Br.H2O in the solid state exhibit dominant, narrow -1/2 <--> +1/2 central transition resonances and associated +/-1/2 <--> +/-3/2 satellite transition resonances which are characteristic of first-order quadrupole broadened systems, while associated high-resolution 65Cu MAS NMR data provide accurate measurement of the 65Cu isotropic chemical shifts. Both approaches provide complete data on the quadrupole and chemical shift interactions which contribute to these spectra. Far-IR spectra of products of reactions involving a range of CuX : t-BuNC ratios reveal the existence of 1 : 1.5 adducts for X = Br, I. Metal-carbon and metal-halogen bands are assigned in the far-IR spectra, which indicate a binuclear double halogen-bridged structure for the 1 : 1.5 complexes.
Sigma-Aldrich
tert-Butyl isocyanide, 98%
Pricing and availability is not currently available.