Merck
  • The first soluble conjugated poly(2,6-anthrylene): synthesis and properties.
The first soluble conjugated poly(2,6-anthrylene): synthesis and properties.
Chemical communications (Cambridge, England) (2008-02-20)
Weibin Cui, Yubo Wu, Hongkun Tian, Yanhou Geng, Fosong Wang
ABSTRACT
The first soluble conjugated poly(2,6-anthrylene) with 9,10-diphenyl-anthracene as the repeating unit is reported; photophysical studies reveal that this polymer represents a novel well-conjugated system.
Product Description
Supelco
9,10-Diphenylanthracene, analytical standard