Merck
  • Development of an immunochromatographic assay for the rapid detection of bromoxynil in water.
Development of an immunochromatographic assay for the rapid detection of bromoxynil in water.
Environmental pollution (Barking, Essex : 1987) (2008-02-08)
Jiang Zhu, Wenchao Chen, Yitong Lu, Guohua Cheng
ABSTRACT
A rapid immunochromatographic one-step strip test was developed to specifically determine bromoxynil in surface and drinking water by competitive inhibition with the nano colloidal gold-conjugated monoclonal antibody (mAb). Bromoxynil standard samples of 0.01-10 mg L(-1) in water were tested by this method and the visual limit was 0.06 mg L(-1). The assay only required 5 min and one-step by dispensing a drop of sample solution onto a strip. Parallel analysis of water samples with bromoxynil showed comparable results from one-step strip test and ELISA. Therefore, the one-step strip test is very useful as a screening method for qualitative detection of bromoxynil in water.
Product Description
Supelco
Bromoxynil, PESTANAL®, analytical standard
Pricing and availability is not currently available.