Merck
  • Sequence-specific recognition of double-stranded DNA by synthetic minor groove binder conjugates. toward the construction of artificial site-specific deoxyribonucleases.
Sequence-specific recognition of double-stranded DNA by synthetic minor groove binder conjugates. toward the construction of artificial site-specific deoxyribonucleases.
Nucleosides, nucleotides & nucleic acids (2007-12-11)
Alexandre S Boutorine, Ludovic Halby, Philippe Simon, Loïc Perrouault, Carine Giovannangeli, Georgy V Gursky, Anna N Surovaya, Sergueï L Grokhovsky, Vladimir A Ryabinin, Alexandre N Sinyakov
ABSTRACT
Bis-conjugates of hairpin N-methylpyrrole/N-methylimidazole oligocarboxamide minor groove binders (MGB) possessing enhanced affinity and sequence-specificity for dsDNA were synthesized. Two hairpin MGBs were connected by their N-termini via an aminodiacetate linker. The binding of bis-MGB conjugates to the target DNA was studied by gel mobility retardation, footprinting, and circular dichroism; their affinity and binding mode in the DNA minor groove were determined. In order to functionalize the bis-MGB conjugates, DNA-cleaving agents such as phenanthroline or bipyridine were attached. Effective site-specific cleavage of target DNA in the presence of Cu(2+) ions was observed.
Sigma-Aldrich
N-Methylpyrrole, 99%
Pricing and availability is not currently available.