Merck
  • Preconcentration of trace arsenite and arsenate with titanium dioxide nanoparticles and subsequent determination by silver diethyldithiocarbamate spectrophotometric method.
Preconcentration of trace arsenite and arsenate with titanium dioxide nanoparticles and subsequent determination by silver diethyldithiocarbamate spectrophotometric method.
Water environment research : a research publication of the Water Environment Federation (2007-10-04)
Yabing Xiao, Jie Ling, Shahua Qian, Anqing Lin, Wenjie Zheng, Weiya Xu, Yuxuan Luo, Man Zhang
ABSTRACT
A novel method of preconcentration of trace arsenite and arsenate by using titanium dioxide nanoparticles as adsorbent was described. The concentrations of preconcentrated arsenite and arsenate were determined by a silver diethyldithiocarbamate spectrophotometric method without desorption. Batch adsorption experiments were carried out as a function of the pH, contact time, amount of titanium dioxide nanoparticles, and solution volume. In the pH range 5 to 6, adsorption rates of arsenite and arsenate were higher than 98%. The calibration coefficient was 0.9991, and the linear range was 0 to 100 microg/L. The developed method was precise, with the relative standard deviation <5% at concentration level of 10 microg/L, with a detection limit (3sigma, n=6) of 0.44 microg/L. The accuracy of the method for total arsenic was validated by standard reference materials (SRM 3103a) (National Institute of Standards and Technology, Gaithersburg, Maryland). The method was also applied to the analysis of arsenite and arsenate in natural water samples to verify the accuracy. The recovery values remained in a narrow range, from 95 to 103%.
Sigma-Aldrich
Silver diethyldithiocarbamate, ACS reagent, for the determination of As, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Silver diethyldithiocarbamate, ACS reagent, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Silver diethyldithiocarbamate
Pricing and availability is not currently available.