Merck
  • Monitoring the efficacy and metabolism of phenylcarbamates in sugar beet and black nightshade by chlorophyll fluorescence parameters.
Monitoring the efficacy and metabolism of phenylcarbamates in sugar beet and black nightshade by chlorophyll fluorescence parameters.
Pest management science (2007-04-18)
Majid Abbaspoor, Jens C Streibig
ABSTRACT
Desmedipham, phenmedipham and a 50% mixture of the two decreased the maximum quantum efficiency of photosystem II (F(v)/F(m)) and the relative changes at the J step (F(vj)) immediately after spraying in both sugar beet and black nightshade grown in the greenhouse. Sugar beet recovered more rapidly from phenmedipham and the mixture than from desmedipham. Desmedipham and the mixture irreversibly affected F(v)/F(m) and F(vj) in black nightshade at much lower doses than in sugar beet. Black nightshade recovered from phenmedipham injury at the highest dose in the first experiment (120 g AI ha(-1)) but not in the second experiment (500 g AI ha(-1)). The dry matter dose-response relationships and the energy pipeline presentation confirmed the same trend. There was a relatively good correlation between F(vj) taken 1 day after spraying and dry matter taken 2 or 3 weeks after spraying. The differential speed of herbicide metabolism between weed and crop plays an important role in herbicide selectivity and can be studied by using appropriate chlorophyll a fluorescence parameters.
Product Description
Supelco
Phenmedipham, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Desmedipham, PESTANAL®, analytical standard
Pricing and availability is not currently available.