Merck
  • Development of a monoclonal antibody-based enzyme-linked immunosorbent assay for the herbicide chlorimuron-ethyl.
Development of a monoclonal antibody-based enzyme-linked immunosorbent assay for the herbicide chlorimuron-ethyl.
Journal of agricultural and food chemistry (2006-07-06)
Jing Zhao, Guo-Xiang Yi, Su-Ping He, Bao-Min Wang, Cai-Xia Yu, Gang Li, Zhi-Xi Zhai, Zhao-Hu Li, Qing X Li
ABSTRACT
Hybridomas secreting a monoclonal antibody (mAb) against the herbicide chlorimuron-ethyl (CE) were produced by fusing the mouse myeloma cell line (SP2/0) with splenocytes from a mouse immunized against the conjugate of the sulfonamide moiety of CE and bovine serum albumin (BSA). The mAb, designated 1F5C5A10, had very weak affinity with metsulfuron, ethametsulfuron, pyrazosulfuron, bensulfuron, and chlorsulfuron. Two mAb-based indirect competitive enzyme-linked immunosorbent assays (icELISA) were developed. A conventional icELISA (icELISA-I) showed a concentration of half-maximum inhibition (IC(50)) of 11.6 ng/mL with a dynamic range of 1.6-84 ng/mL. A simplified icELISA (icELISA-II) had an IC(50) of 28.7 ng/mL and a dynamic range of 2.2-372 ng/mL. The two assays were tested on spiked water and soil samples. CE (1-500 ng/mL) fortified in water samples could be analyzed directly without any sample preparation by both immunoassays with an average recovery between 74 and 114%. icELISA-II, but not icELISA-I, was able to accurately analyze the herbicide residues in the crude soil extracts with recoveries between 99 and 129% without obvious matrix effects due to its lesser amount of sample used. In contrast to icELISA-I, icELISA-II is more convenient, whereas it consumes more reagents of coating antigen and goat anti-mouse IgG-peroxidase.
Product Description
Supelco
Chlorimuron ethyl, PESTANAL®, analytical standard
Pricing and availability is not currently available.