Merck
  • Conformational studies of proline-, thiaproline- and dimethylsilaproline-containing diketopiperazines.
Conformational studies of proline-, thiaproline- and dimethylsilaproline-containing diketopiperazines.
Journal of peptide science : an official publication of the European Peptide Society (2006-06-21)
Florine Cavelier, Damien Marchand, Patrick Mbassi, Jean Martinez, Michel Marraud
ABSTRACT
As proline plays an important role in biologically active peptides, many analogues of this residue have been developed to modulate the proportion of cis and trans conformers. A correlation between the pyrrolidine ring shape and structural properties of proline has been established. Diketopiperazine (DKP) is the model of choice to study the influence of the proline ring modification. In this contribution, cyclo(Gly-Pro) and two analogues cyclo(Sip-Pro) and cyclo(Thz-Pro) have been studied with proton NMR. We showed that both analogues with heteroatoms in gamma position, silicon and sulfur respectively, display a more rigid five-member ring. The usual flexibility of proline ring is restrained in both cases and only the two C(beta)-exo and C(beta)-endo conformations are observed.
Sigma-Aldrich
L-4-Thiazolidinecarboxylic acid, 98%
Pricing and availability is not currently available.