Merck
  • Recent advances in the solid-phase combinatorial synthetic strategies for the quinoxaline, quinazoline and benzimidazole based privileged structures.
Recent advances in the solid-phase combinatorial synthetic strategies for the quinoxaline, quinazoline and benzimidazole based privileged structures.
Mini reviews in medicinal chemistry (2006-02-07)
Ahmed Kamal, K Laxma Reddy, V Devaiah, N Shankaraiah, M V Rao
ABSTRACT
Quinoxaline, quinazoline and benzimidazole based templates have been synthesized on solid-support employing different methodologies. This review enlightens academic and industrial examples of combinatorial synthesis for this type of heterocycles that appeared in the literature in the last decade. Hence, some of the important synthetic strategies for the generation of quinoxaline, quinazoline and benzimidazole based privileged structures, and the important biological activities for these heterocycles have been highlighted. Further, benzothiadiazinone, thioxoquinazolinone, cinnoline and indazole are also examined in this review.
Sigma-Aldrich
4-(2-Keto-1-benzimidazolinyl)piperidine, 98%
Pricing and availability is not currently available.