Merck
  • Analysis of the antinociceptive effect of the flavonoid myricitrin: evidence for a role of the L-arginine-nitric oxide and protein kinase C pathways.
Analysis of the antinociceptive effect of the flavonoid myricitrin: evidence for a role of the L-arginine-nitric oxide and protein kinase C pathways.
The Journal of pharmacology and experimental therapeutics (2005-11-02)
Flavia Carla Meotti, Ana Paula Luiz, Moacir Geraldo Pizzolatti, Cândida A L Kassuya, João B Calixto, Adair R S Santos
ABSTRACT
The present study investigated the antinociceptive effects of the flavonoid myricitrin in chemical behavioral models of pain in mice and rats. Myricitrin given by i.p. or p.o. routes produced dose-related antinociception when assessed on acetic acid-induced visceral pain in mice. In addition, the i.p. administration of myricitrin exhibited significant inhibition of the neurogenic pain induced by intraplantar (i.pl.) injection of capsaicin. Like-wise, myricitrin given by i.p. route reduced the nociception produced by i.pl. injection of glutamate and phorbol myristate acetate (PMA). Western blot analysis revealed that myricitrin treatment fully prevented the protein kinase C (PKC) alpha and PKCepsilon activation by PMA in mice hind paws. Myricitrin given i.p. also inhibited the mechanical hyperalgesia induced by bradykinin, without affecting similar responses caused by epinephrine and prostaglandin E(2). The antinociception caused by myricitrin in the acetic acid test was significantly attenuated by i.p. treatment of mice with the nitric oxide precursor, L-arginine. In contrast, myricitrin antinociception was not affected by naloxone (opioid receptor antagonist) or neonatal pretreatment of mice with capsaicin and myricitrin antinociceptive effects is not related to muscle relaxant or sedative action. Together, these results indicate that myricitrin produces pronounced antinociception against chemical and mechanical models of pain in rodents. The mechanisms involved in their actions are not completely understood but seem to involve an interaction with nitric oxide-L-arginine and protein kinase C pathways.
Product Description
Supelco
Myricitrin, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Myricitrin, ≥99.0% (HPLC)
Pricing and availability is not currently available.