Merck
  • Uncatalyzed reaction of silyl ketene acetals with oxalyl chloride: a straightforward preparation of symmetrical pulvinic acids.
Uncatalyzed reaction of silyl ketene acetals with oxalyl chloride: a straightforward preparation of symmetrical pulvinic acids.
The Journal of organic chemistry (2005-02-12)
Benoît Heurtaux, Claude Lion, Thierry Le Gall, Charles Mioskowski
ABSTRACT
[reaction: see text] Several natural pulvinic acids were synthesized. Silyl ketene acetals derived from methyl arylacetates (4 equiv) reacted with oxalyl chloride at -78 degrees C, without the need of adding a catalyst. After treatment of the crude diketones with DBU and acidification with hydrochloric acid, symmetrical pulvinic acids methyl esters were obtained. Saponification of the methyl esters afforded the corresponding pulvinic acids in 60-70% overall yields from oxalyl chloride.
Sigma-Aldrich
Oxalyl chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalyl chloride, reagent grade, 98%