Merck
  • Early reperfusion levels of Na(+) and Ca(2+) are strongly associated with postischemic functional recovery but are disassociated from K(ATP) channel-induced cardioprotection.
Early reperfusion levels of Na(+) and Ca(2+) are strongly associated with postischemic functional recovery but are disassociated from K(ATP) channel-induced cardioprotection.
Journal of molecular and cellular cardiology (2004-07-28)
Eiichi Takayama, Ling-Ling Guo, Stanley B Digerness, Martin M Pike
ABSTRACT
We previously demonstrated that pinacidil does not affect Na(+)(i) accumulation, cellular energy depletion, or acidosis during myocardial ischemia, but dramatically improves the cationic/energetic status during reperfusion. We investigated the role of this latter effect in K(ATP) channel-induced cardioprotection. Employing (23)Na and (31)P nuclear magnetic resonance spectroscopy with perfused rat hearts, reperfusion Na(+)(i) was altered with brief infusions of ouabain and/or RbCl to transiently decrease or increase Na(+)/K(+) ATPase activity. The increases and decreases in functional recovery (%LVDP-R) with pinacidil or ouabain, respectively, were largely unaltered by each other's presence. Early reperfusion Na(+)(i) and cellular energy were greatly altered by ouabain and indicated linear relationships with %LVDP-R. Pinacidil shifted these relationships to higher %LVDP-R. Increasing early reperfusion Na(+)(i) decreased %LVDP-R but did not diminish pinacidil's capacity to improve %LVDP-R. Approximately 75% and 45% of the pinacidil-induced improvements in %LVDP-R, could be disassociated from early reperfusion Na(+)(i) and cellular energy, respectively. Both pinacidil and RbCl infusion blunted ouabain's elevation of reperfusion Na(+)(i), but RbCl did not improve %LVDP-R. Atomic absorption tissue Ca(2+) measurements indicated that pinacidil reduced late reperfusion Ca(2+) uptake, but did not reduce early reperfusion Ca(2+), and its beneficial effects were resistant to ouabain-induced early reperfusion Ca(2+) increases. In conclusion, K(ATP) channel-induced cardioprotection does not require moderation of Na(+)(i) accumulation, cellular energy depletion, or acidosis during ischemia. K(ATP) channel-induced cardioprotection is largely independent of the accelerated reperfusion Na(+)(i) recovery it induces and does not require early reperfusion reductions of tissue Ca(2+). A larger role for early reperfusion cellular energy cannot be excluded.
Sigma-Aldrich
Rubidium chloride, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Rubidium chloride, 99.8% trace metals basis
Sigma-Aldrich
Rubidium chloride, 99.95% trace metals basis
Sigma-Aldrich
Rubidium chloride, ReagentPlus®, ≥99.0% (metals basis)