Merck
Control of Escherichia coli O157:H7 with sodium metasilicate.
Journal of food protection (2004-07-24)
George H Weber, Judy K O'Brien, Fredric G Bender
ABSTRACT
Three intervention strategies-trisodium phosphate, lactic acid, and sodium metasilicate--were examined for their in vitro antimicrobial activities in water at room temperature against a three-strain cocktail of Escherichia coli O157:H7 and a three-strain cocktail of "generic" E. coli. Both initial inhibition and recovery of injured cells were monitored. When 3.0% (wt/wt) lactic acid, pH 2.4, was inoculated with E. coli O157:H7 (approximately 6 log CFU/ml), viable microorganisms were recovered after a 20-min exposure to the acid. After 20 min in 1.0% (wt/wt) trisodium phosphate, pH 12.0, no viable E. coli O157:H7 microorganisms were detected. Exposure of E. coli O157:H7 to sodium metasilicate (5 to 10 s) at concentrations as low as 0.6%, pH 12.1, resulted in 100% inhibition with no recoverable E. coli O157:H7. No difference in inhibition profiles was detected between the E. coli O157:H7 and generic strains, suggesting that nonpathogenic strains may be used for in-plant sodium metasilicate studies.
Sigma-Aldrich
Sodium metasilicate
Pricing and availability is not currently available.