Merck
  • Inhibition of macrophage development and foreign body giant cell formation by hydrophilic interpenetrating polymer network.
Inhibition of macrophage development and foreign body giant cell formation by hydrophilic interpenetrating polymer network.
Journal of biomedical materials research. Part A (2004-05-27)
Terry O Collier, James M Anderson, William G Brodbeck, Thomas Barber, Kevin E Healy
ABSTRACT
The ability of monocytes to adhere, differentiate into macrophages, and fuse to form foreign body giant cells (FBGCs) on an implanted material surface is a critical step toward biomaterial degradation. Novel homogeneous surfaces were utilized to mediate adhesion. These surfaces consisted of N-(2 aminoethyl)-3-aminopropyltrimethoxysilane (EDS) and an interpenetrating polymer network (IPN) of polyacrylamide and poly(ethylene glycol). These surfaces were designed to control cell adhesion and morphology and mediate cell differentiation, activation, metabolic ability, and apoptosis, resulting in a reduced or controlled inflammatory response. The EDS surface promotes cell adhesion and the IPN minimizes protein adsorption and subsequent cell adhesion. Both surfaces had similar cellular adhesion rates at each respective time point. However, the adherent macrophage morphology was similar at 2 h and day 3, and at days 7 and 10 adherent macrophages on the EDS surface formed FBGCs (46% at day 7 and 40% at day 10). Adherent cells on the IPN surface did not form FBGCs but instead formed monocyte aggregates (73% of adherent cells formed aggregates at day 7 and 63% at day 10). It is indicated that the two surface chemistries differentially controlled monocyte differentiation into macrophages and subsequent macrophage fusion to form FBGCs.
Sigma-Aldrich
[3-(2-Aminoethylamino)propyl]trimethoxysilane, technical grade, ≥80%
Sigma-Aldrich
N-[3-(Trimethoxysilyl)propyl]ethylenediamine, 97%