Merck
  • Biosynthetic origin of the rare nitroaryl moiety of the polyketide antibiotic aureothin: involvement of an unprecedented N-oxygenase.
Biosynthetic origin of the rare nitroaryl moiety of the polyketide antibiotic aureothin: involvement of an unprecedented N-oxygenase.
Journal of the American Chemical Society (2004-03-25)
Jing He, Christian Hertweck
ABSTRACT
The antibiotic aureothin is a rare natural nitroaromatic compound produced by Streptomyces thioluteus. By labeling experiments, we demonstrate for the first time that p-nitrobenzoate (PNBA) serves as a polyketide synthase starter unit. Cloning, heterologous expression, and inactivation experiments reveal that this unusual primer is synthesized from p-aminobenzoate (PABA) by means of AurF, a novel N-oxygenase.
Sigma-Aldrich
4-Nitrobenzoic acid, 98%
Sigma-Aldrich
4-Nitrobenzoic acid, purum, ≥98.0% (HPLC)