Merck
  • Air oxidation of d-limonene (the citrus solvent) creates potent allergens.
Air oxidation of d-limonene (the citrus solvent) creates potent allergens.
Contact dermatitis (1992-05-01)
A T Karlberg, K Magnusson, U Nilsson
ABSTRACT
Products containing as much as 95% of d-limonene are used for, e.g., degreasing metal before industrial painting and for cleaning assemblies. Experimental studies on the sensitizing potential of limonene show diverging results. In a previous study, we found that the sensitizing potential of d-limonene increased with prolonged air exposure. The aim of this study was to make further chemical analyses, to identify compounds formed by air exposure of d-limonene and to study their allergenic potential. d-limonene was found to be a sensitizer after prolonged exposure to air according to 2 Freund's complete adjuvant test (FCAT) experiments and 1 guinea pig maximization test (GPMT) study. No significant response was obtained to d-limonene not air exposed, even if the animals were sensitized to oxidized d-limonene. 5 main oxidation products of d-limonene were identified. (R)-(-)-carvone and a mixture of cis and trans isomers of (+)-limonene oxide were found to be potent sensitizers, while no significant reactions were obtained in the animals induced with a mixture of cis and trans isomers of (-)-carveol. It can be concluded that air oxidation of d-limonene is essential for its sensitizing potential, and that potent allergens are created.
Sigma-Aldrich
(−)-Carveol, mixture of isomers, 97%
Sigma-Aldrich
L-Carveol, mixture of cis and trans, ≥95%, FG