MilliporeSigma
  • Two children with bromate intoxication due to ingestion of the second preparation for permanent hair waving.
Two children with bromate intoxication due to ingestion of the second preparation for permanent hair waving.
Acta paediatrica Japonica : Overseas edition (1992-12-01)
T Watanabe, T Abe, M Satoh, Y Oda, T Takada, T Yanagihara
ABSTRACT
We report two children who suffered from sodium bromate intoxication due to ingestion of the second preparation for permanent hair waving (the second permanent preparation). One child suffered from gastrointestinal symptoms only. The other exhibited slight acute renal insufficiency. Results of the histological examination of the kidney in the sick child with acute renal insufficiency showed sporadic epithelial separation of the proximal tubuli under light microscopy. In addition, we could demonstrate more clearly epithelial separation and unbroken tubular basement membranes under electron microscopy (EM). To our knowledge, this is the first report of EM findings in this disease. The mechanism of epithelial injuries by bromate is not clear.
Product Description
Supelco
Sodium bromate, puriss. p.a., ≥99.5% (RT)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$59.50
Sigma-Aldrich
Sodium bromate, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$59.50