Merck
  • Mild synthesis of disaccharidic 2,3-enopyranosyl cyanides and 2-C-2-deoxy pyranosyl cyanides with Hg(CN)(2)/HgBr(2)/TMSCN.
Mild synthesis of disaccharidic 2,3-enopyranosyl cyanides and 2-C-2-deoxy pyranosyl cyanides with Hg(CN)(2)/HgBr(2)/TMSCN.
The Journal of organic chemistry (2002-10-26)
Andreas H Franz, YiQiu Wei, Vyacheslav V Samoshin, Paul H Gross
ABSTRACT
Lewis acid-catalyzed dimerization of mono- and disaccharidic per-O-acetylated glycals gave di- and tetrasaccharidic O-acetylated C-glycosides, respectively. 2,3-Enopyranosyl cyanides were obtained from per-O-acetylated glycals by a new, mild anomeric S(N)'-acetoxy displacement with Hg(CN)(2)/HgBr(2)/TMSCN. Per-O-acetylated 2-C-2-deoxy-pyranoses were converted into pyranosyl cyanides by the same reagent. An unprecedented acetic acid elimination from dimers with D-galacto- and L-fuco-configurations accompanied the S(N)-displacement under those conditions. A new set of (1)H NMR coupling constants for 2,3-enopyranosyl systems was used for configurational assignment of complicated tetrasaccharide mimics.
Sigma-Aldrich
Tri-O-acetyl-D-glucal, 98%
Pricing and availability is not currently available.