Merck
  • New supramolecular organization for a glycoluril: chiral hydrogen-bonded ribbons.
New supramolecular organization for a glycoluril: chiral hydrogen-bonded ribbons.
Chemical communications (Cambridge, England) (2002-10-26)
Darren W Johnson, Liam C Palmer, Fraser Hof, Peter M Iovine, Julius Rebek
ABSTRACT
A series of substituted glycoluril molecules exhibits a substantial twist of the fused five-membered rings and assembles exclusively chiral hydrogen-bonded ribbons in the solid-state.
Sigma-Aldrich
Glycoluril, 97%