Merck
  • Parameters influencing protein extraction for whole broths in detergent based aqueous two-phase systems.
Parameters influencing protein extraction for whole broths in detergent based aqueous two-phase systems.
Bioseparation (2002-09-18)
K Selber, A Collén, T Hyytiä, M Penttilä, F Tjerneld, M R Kula
ABSTRACT
The parameters important for an optimisation of cloud point extraction in technical scale were investigated using a genetically engineered fusion protein derived from endoglucanase I expressed in Trichoderma reesei and the nonionic polyoxyethylene Agrimul NRE 1205. The key parameters are temperature, detergent concentration, and additional salts. These parameters are interdependent, thus there is an optimum in the partition coefficient with respect to detergent concentration and a maximum for the partition coefficient and the yield with respect to temperature. These results were confirmed for the detergent C12E5 to demonstrate that these optima are due to the nature of polyoxyethylenes. Cloud point extraction was found to be only slightly affected by pH. In the case studied extraction of whole broth is favourable for a high yield and partition coefficient, since fusion protein adhering to the cells can be solubilized. However some loss of detergent which remains in the fungal biomass was observed.
Sigma-Aldrich
Pentaethylene glycol monododecyl ether, BioXtra, ≥98.0% (GC)
Pricing and availability is not currently available.