Merck
  • Anaerobic biodegradability of alkylphenols and fuel oxygenates in the presence of alternative electron acceptors.
Anaerobic biodegradability of alkylphenols and fuel oxygenates in the presence of alternative electron acceptors.
Applied microbiology and biotechnology (2000-12-29)
L Puig-Grajales, N G Tan, F van der Zee, E Razo-Flores, J A Field
ABSTRACT
Alkylphenols and fuel oxygenates are important environmental pollutants produced by the petrochemical industry. A batch biodegradability test was conducted with selected ortho-substituted alkylphenols (2-cresol, 2,6-dimethylphenol and 2-ethylphenol), fuel oxygenates (methyl tert-butyl ether, ethyl tert-butyl ether and tert-amylmethyl ether) and tert-butyl alcohol (TBA) as model compounds. The ortho-substituted alkylphenols were not biodegraded after 100 days of incubation under methanogenic, sulfate-, or nitrate-reducing conditions. However, biodegradation of 2-cresol and 2-ethylphenol (150 mg l(-1)) was observed in the presence of Mn (IV) as electron acceptor. The biodegradation of these two compounds took place in less than 15 days and more than 90% removal was observed for both compounds. Mineralization was indicated since no UV-absorbing metabolites accumulated after 23 days of incubation. These alkylphenols were also slowly chemically oxidized by Mn (IV). No biodegradation of fuel oxygenates or TBA (1 g l(-1)) was observed after 80 or more days of incubation under methanogenic, Fe (III)-, or Mn (IV)-reducing conditions, suggesting that these compounds are recalcitrant under anaerobic conditions. The fuel oxygenates caused no toxicity towards acetoclastic methanogens activity in anaerobic granular sludge.
Sigma-Aldrich
2,6-Dimethylphenol, 99%
Pricing and availability is not currently available.
Supelco
2,6-Dimethylphenol, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
2,6-Dimethylphenol, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
2,6-Xylenol, ≥99%, FG
Pricing and availability is not currently available.