Merck
  • Crystallization and preliminary X-ray crystallographic analysis of NADPH: azodicarbonyl/quinone oxidoreductase, a plant zeta-crystallin.
Crystallization and preliminary X-ray crystallographic analysis of NADPH: azodicarbonyl/quinone oxidoreductase, a plant zeta-crystallin.
Biochimica et biophysica acta (2000-09-27)
J Mano, H Yoon, K Asada, E Babiychuk, D Inzé, B Mikami
ABSTRACT
Arabidopsis thaliana P1 protein was crystallized using the hanging drop vapor-diffusion method in 0.1 M piperazine-1, 4-bis(2-ethanesulfonic acid) buffer, containing 14% polyethylene glycol 6000 and 0.2 M magnesium acetate at pH 6.5 and 20 degrees C. The crystals are orthorhombic and belong to the space group P2(1)2(1)2(1) with unit cell dimensions of a=49.8, b=122.4 and c=149. 9 A. The diffraction data up to 2.9 A were collected by a multiwire area detector.
Sigma-Aldrich
PIPES, BioXtra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
PIPES, ≥99% (titration)
Sigma-Aldrich
PIPES, BioPerformance Certified, suitable for cell culture
Sigma-Aldrich
PIPES, BioXtra, ≥99% (titration)
Sigma-Aldrich
PIPES sodium salt, ≥99% (titration)