Merck
  • Development of a bioconversion process for production of cis-1S,2R-indandiol from indene by recombinant Escherichia coli constructs.
Development of a bioconversion process for production of cis-1S,2R-indandiol from indene by recombinant Escherichia coli constructs.
Applied microbiology and biotechnology (1999-07-03)
J Reddy, C Lee, M Neeper, R Greasham, J Zhang
ABSTRACT
Recombinant Escherichia coli cells expressing the toluene dioxygenase (TDO) genes from Pseudomonas putida convert indene to cis-1S,2R-indandiol, a potentially important intermediate for the chemical synthesis of the HIV-1 protease inhibitor, Crixivan. A bioconversion process was developed through optimization of medium composition and reaction conditions at the shake-flask and 23-1 fermentor scales. A cis-1,2-indandiol productivity of approx. 1000 mg/l was achieved with construct TDO123, which represents a 50-fold increase over the initial titer. Varying the bioconversion conditions did not change the enantiomeric excess (e.e.) for the 1S,2R enantiomer from about 30%, suggesting that toluene dioxygenase intrinsically converts indene to 1S,2R- and 1R,2S-indandiols at a ratio of 2:1. Further inclusion of the Pseudomonas dehydrogenase gene in construct D160-1 led to the production of chirally pure cis-1S,2R-indandiol (e.e. > 99%) as a result of the selective degradation of the 1R,2S enantiomer, with the overall yield (650 mg/l) proportionally reduced. A single stage process was developed for D160-1 and scaled up to the 23-1 fermentor, achieving a cis-1S,2R-indandiol titer of 1200 mg/l.
Sigma-Aldrich
(R)-(−)-1-Indanol, 99%
Pricing and availability is not currently available.