Merck
  • Analysis of sodium adduct paeoniflorin, albiflorin and their derivatives by (+)ESI-MSn, DFT calculations and computer-aided mass spectrometry analysis program.
Analysis of sodium adduct paeoniflorin, albiflorin and their derivatives by (+)ESI-MSn, DFT calculations and computer-aided mass spectrometry analysis program.
Journal of mass spectrometry : JMS (2007-01-03)
Rui Li, Xiaoling Wang, Yan Zhou, Min Cai, Lisheng Ding
ABSTRACT
In order to analyze paeoniflorin, albiflorin and their derivatives (PADs) in Paeonia Lactiflora rapidly and effectively, (+)ESI-MS(n) experiments were conducted, from which two diagnostic fragment patterns were acquired. Meanwhile, the dehydration ability of aglycones of PADs was obtained by calculating their activation energy using density functional theory, through which the unique dehydration phenomenon of benzoylalbiflorin, compared with benzoylpaeoniflorin, was interpreted. In addition, a computer-aided mass spectrometry analysis program was developed to facilitate the analysis of the unknown compound by suggesting the possible structure of the analyte.
Sigma-Aldrich
Albiflorin, ≥95% (LC/MS-ELSD)
Pricing and availability is not currently available.