Merck
Poly[diacetonitrile-[μ(3)-difluoro-(oxalato)borato]sodium].
Acta crystallographica. Section E, Structure reports online (2011-07-15)
Joshua L Allen, Paul D Boyle, Wesley A Henderson
ABSTRACT
The title compound, [Na(C(2)BF(2)O(4))(CH(3)CN)(2)](n), forms infinite two-dimensional layers running parallel to (010). The layers lie across crystallographic mirror planes at y = 1/4 and 3/4. The Na, B and two F atoms reside on these mirror planes. The Na(+) cations are six-coordinate. Two equatorial coordination positions are occupied by acetonitrile mol-ecules. The other two equatorial coordination sites are occupied by the chelating O atoms from the difluoro-(oxalato)borate anion (DFOB(-)). The axial coordination sites are occupied by two F atoms from two different DFOB(-) anions.
Product Description
Supelco
Sodium oxalate, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Sodium oxalate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, for HPLC, for UV, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Sodium oxalate, puriss. p.a., ACS reagent, ≥99.5% (RT)
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Sodium oxalate, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Sodium oxalate, BioXtra
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis