Merck
  • Pharmacological characterization of 1-aminoindan-1,5-dicarboxylic acid, a potent mGluR1 antagonist.
Pharmacological characterization of 1-aminoindan-1,5-dicarboxylic acid, a potent mGluR1 antagonist.
The Journal of pharmacology and experimental therapeutics (1997-05-01)
F Moroni, G Lombardi, C Thomsen, P Leonardi, S Attucci, F Peruginelli, S A Torregrossa, D E Pellegrini-Giampietro, R Luneia, R Pellicciari
ABSTRACT
We examined the pharmacological profile of 1-aminoindan-1,5-dicarboxylic acid (AIDA), a rigid (carboxyphenyl)glycine derivative acting on metabotropic glutamate receptors (mGluRs). In cells transfected with mGluR1a, AIDA competitively antagonized the stimulatory responses of glutamate and (1S,3R)-1-aminocyclopentane-1,3-dicarboxylic acid [(1S,3R)-ACPD] on phosphoinositide hydrolysis (pA2 = 4.21). In cells transfected with mGluR5a, AIDA displayed a much weaker antagonist effect. In transfected cells expressing mGluR2, AIDA (< or = 1 mM) did not affect the inhibition of forskolin-stimulated adenylate cyclase activity induced by (1S,3R)-ACPD, but at large concentrations, it displayed a modest agonist activity. In rat hippocampal or striatal slices, AIDA (0.1-1 mM) reduced the effects of (1S,3R)-ACPD on phospholipase C but not on adenylate cyclase responses, whereas (+)-alpha-methyl-4-carboxyphenylglycine (0.3-1 mM) was an antagonist on both transduction systems. In addition, AIDA (0.3-1 mM) had no effect on mGluRs coupled to phospholipase D, whereas (+)-alpha-methyl-4-carboxy-phenylglycine (0.5-1 mM) acted as an agonist with low intrinsic activity. In rat cortical slices, AIDA antagonized the stimulatory (mGluR1-mediated) effect of (1S,3R)-ACPD on the depolarization-induced outflow of D-[3H]aspartate, disclosing an inhibitory effect ascribable to (1S,3R)-ACPD activating mGluR2 and/or mGluR4. Finally, mice treated with AIDA (0.1-10 nmol i.c.v.) had an increased pain threshold and difficulties in initiating a normal ambulatory behavior. Taken together, these data suggest that AIDA is a potent, selective and competitive mGluR1 a antagonist.
Sigma-Aldrich
AIDA, solid
Pricing and availability is not currently available.