Merck
  • Molecular cloning, purification and characterization of thermostable beta-1,3-1,4 glucanase from Bacillus subtilis A8-8.
Molecular cloning, purification and characterization of thermostable beta-1,3-1,4 glucanase from Bacillus subtilis A8-8.
Indian journal of biochemistry & biophysics (2010-12-24)
Youn-Ju Jung, Yong-Seok Lee, In-Hye Park, M Subhosh Chandra, Keun-Ki Kim, Yong-Lark Choi
ABSTRACT
A gene encoding a beta-1,3-1,4-glucanase (CelA) belonging to family 5 of glycoside hydrolases was cloned and sequenced from the Bacillus subtilis A8-8. The open-reading-frame of celA comprised 1499 base pairs and the enzyme was composed of 500 amino acids with a molecular mass of 55 kDa. The recombinant beta-1,3-1,4 glucanase was purified by GST-fusion purification system. The pH and temperature optima of the enzyme were 8.0 and 60 degrees C, respectively. The enzyme was stable within pH 6.0-9.0. It was stable up to 60 degrees C and retained 30% of its original activity at 70 degrees C for 60 min. It hydrolyzed lichenan, CMC, xylan, laminarin, avicel and pNPC, but was inactive towards cellobiose. The enzyme activity was markedly activated by Co2+ and Mn2+, but was strongly inactivated by Fe3+. The truncated gene, devoid of cellulose-binding domain (CBD) showed 60% of activity and bound to avicel.
Sigma-Aldrich
β-Glucanase from Aspergillus niger, powder, dark brown, ~1 U/mg
Sigma-Aldrich
β-Glucanase from Trichoderma longibrachiatum