Merck
  • [Specific features of the anticonvulsant effect of memantine in mice intoxicated with a model organic phosphate].
[Specific features of the anticonvulsant effect of memantine in mice intoxicated with a model organic phosphate].
Eksperimental'naia i klinicheskaia farmakologiia (2013-05-02)
M A Iudin, S V Chepur, V N Bykov, A F Kurpiakova, S N Subbotina, A S Nikiforov, I M Ivanov
ABSTRACT
The effect of memantine administration has been studied on the model of mice poisoning with an anticholinesterase compound. It is established that the memantine action is due to its influence on the cholinesterase activity in the brain, blood plasma, and erythrocytes in addition to its NMDA-blocking action. Memantine promotes oxime-induced erythrocyte enzyme reactivation on the model of mice poisoning with anticholinesterase compound (0.8 LD50).
Sigma-Aldrich
Acetylcholinesterase human, recombinant, expressed in HEK 293 cells, lyophilized powder, ≥1,000 units/mg protein (Lowry)
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type V-S, lyophilized powder, ≥1,000 units/mg protein
Sigma-Aldrich
Acetylcholinesterase from Electrophorus electricus (electric eel), Type VI-S, lyophilized powder, 200-1,000 units/mg protein
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
Acetylcholinesterase from human erythrocytes, buffered aqueous solution, ≥500 units/mg protein (BCA)