Merck
  • A single-molecule analysis reveals morphological targets for cellulase synergy.
A single-molecule analysis reveals morphological targets for cellulase synergy.
Nature chemical biology (2013-04-09)
Jerome M Fox, Phillip Jess, Rakesh B Jambusaria, Genny M Moo, Jan Liphardt, Douglas S Clark, Harvey W Blanch
ABSTRACT
The mechanisms of enzyme activity on solid substrates are not well understood. Unlike enzyme catalysis in aqueous solutions, enzyme activity on surfaces is complicated by adsorption steps and structural heterogeneities that make enzyme-substrate interactions difficult to characterize. Cellulase enzymes, which catalyze the depolymerization of cellulose, show binding specificities for different cellulose surface morphologies, but the influence of these specificities on the activity of multienzyme mixtures has remained unclear. We developed a metric to quantify binding-target arrangements determined by photoactivated localization microscopy, and we used that metric to show that combinations of cellulases designed to bind within similar but nonidentical morphologies can have synergistic activity. This phenomenon cannot be explained with the binary crystalline or amorphous classifications commonly used to characterize cellulase-binding targets. Our results reveal a strategy for improving the activity of cellulolytic mixtures and demonstrate a versatile method for investigating protein organization on heterogeneous surfaces.
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Cellulase from Aspergillus sp., aqueous solution
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid
Supelco
Cellulose, powder, for column chromatography
Supelco
Cellulose, acid washed, powder, for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, acid washed, from spruce, for column chromatography
Sigma-Aldrich
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., BioReagent, suitable for plant cell culture, 3-10 units/mg solid
Sigma-Aldrich
Cellulose, fibers, (medium)
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.