Merck
  • Differential transport of Influenza A neuraminidase signal anchor peptides to the plasma membrane.
Differential transport of Influenza A neuraminidase signal anchor peptides to the plasma membrane.
FEBS letters (2013-03-26)
Andreas Max Ernst, Sonja Zacherl, Alexia Herrmann, Moritz Hacke, Walter Nickel, Felix T Wieland, Britta Brügger
ABSTRACT
Influenza A Neuraminidase is essential for virus release from the cell surface of host cells. Given differential structures of the N-terminal sequences including the transmembrane domains of neuraminidase subtypes, we investigated their contribution to transport and localization of subtypes N1, N2 and N8 to the plasma membrane. We generated consensus sequences from all protein entries available for these subtypes. We found that 40N-terminal the forty N-terminal amino acids are sufficient to confer plasma membrane localization of fusion proteins, albeit with different efficiencies. Strikingly, subtle differences in the primary structure of the part of the transmembrane domain that resides in the exoplasmic leaflet of the membrane have a major impact on transport efficiency, providing a potential target for the inhibition of virus release.
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type X, lyophilized powder, ≥50 units/mg protein (using 4MU-NANA)
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VIII, lyophilized powder, 10-20 units/mg protein (using 4MU-NANA), 3.5-8.0 units/mg protein (mucin)
Sigma-Aldrich
α(2→3) Neuraminidase from Streptococcus pneumoniae, buffered aqueous solution
Sigma-Aldrich
α(2→3,6) Neuraminidase from Clostridium perfringens (C. welchii), recombinant, expressed in E. coli, buffered aqueous solution, ≥250 units/mg protein
Sigma-Aldrich
Neuraminidase Agarose from Clostridium perfringens (C. welchii), Type VI-A, ammonium sulfate suspension
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Suitable for manufacturing of diagnostic kits and reagents, Type V, lyophilized powder
Sigma-Aldrich
Neuraminidase from Clostridium perfringens (C. welchii), Type VI, lyophilized powder, 6-15 units/mg protein (using 4MU-NANA), 2-10 units/mg protein (mucin)
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, ≥1.5 U/mL, specific activity ≥ 1.5U/mg protein
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type III, buffered aqueous solution, 0.2 μm filtered, 1-5 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Neuraminidase from Vibrio cholerae, Type II, buffered aqueous solution, 8-24 units/mg protein (Lowry, using NAN-lactose)
Sigma-Aldrich
α(2→3,6,8,9) Neuraminidase from Arthrobacter ureafaciens, Proteomics Grade, suitable for MALDI-TOF MS