Merck
  • Klotho suppresses tumor progression via inhibiting PI3K/Akt/GSK3β/Snail signaling in renal cell carcinoma.
Klotho suppresses tumor progression via inhibiting PI3K/Akt/GSK3β/Snail signaling in renal cell carcinoma.
Cancer science (2013-02-26)
Yu Zhu, Le Xu, Jianping Zhang, Wenping Xu, Yujun Liu, Hankun Yin, Tao Lv, Huimin An, Li Liu, Hongyong He, Heng Zhang, Jing Liu, Jiejie Xu, Zongming Lin
ABSTRACT
Klotho is an anti-aging protein predominantly expressed in renal tubular epithelial cells. Although Klotho was recently identified as a tumor suppressor gene in a variety of cancers, the potential role and molecular events for Klotho in renal cell carcinoma (RCC) remain obscure. In the present study, immunohistochemical staining in tissue microarrays containing 125 RCC samples showed that intratumoral Klotho levels were negatively correlated with tumor size, TNM stage and nuclear grade. The overall survival rate of RCC patients with high Klotho expression was significantly higher than that of patients with low Klotho expression. Functional analysis after gain and loss of Klotho expression revealed that Klotho blunted epithelial-mesenchymal transition and cellular migration and invasion in RCC. Also, no alteration of α-2,6-sialidase activity was found after Klotho overexpression in RCC. The molecular signals for this phenomenon involved the Klotho-mediated inhibition of PI3K/Akt/GSK3β/Snail pathway. Importantly, compared to localized RCC tissues, advanced RCC tissues exhibited low Klotho expression accompanied with high pAkt and Snail expression. These results indicate Klotho acts as a tumor suppressor by inhibiting PI3K/Akt/GSK3β/Snail signaling, thus suppressing epithelial-mesenchymal transition and tumor migration and invasion during RCC progression. As a result, Klotho might be used as a potential therapy for advanced RCC.
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution