Merck
  • Profiling mRNA of the graying human hair follicle constitutes a promising state-of-the-art tool to assess its aging: an exemplary report.
Profiling mRNA of the graying human hair follicle constitutes a promising state-of-the-art tool to assess its aging: an exemplary report.
The Journal of investigative dermatology (2012-12-14)
Eva M J Peters, Christiane Liezmann, Katharina Spatz, Ute Ungethüm, Ralf-Jürgen Kuban, Maria Daniltchenko, Johannes Kruse, Dominik Imfeld, Burghard F Klapp, Remo Campiche
ABSTRACT
Determining hitherto uninvestigated and safe targets to halt the aging process is important in our aging society. Graying is a hallmark of the aging process and may be used to identify aging tissue for comparative analysis. Here we analyzed differential gene expressions between pigmented, gray, and white human scalp skin hair follicles (HFs) from identical donors. Forming intersections between five donors identified 194/192 downregulated and 186/177 upregulated genes in gray/white HFs. These included melanogenesis (tyrosinase; tyrosinase-related protein 1)- and melanosome structure (Melan-A; Pmel17)-associated genes and regulation of melanocyte relevant tyrosine kinases. Alongside these expected changes, regulated genes included nonmelanocyte-related genes associated with aging as well as nonaging-related genes associated with melanocytes. Intriguingly, among them, genes associated with energy metabolism (i.e., glutaminase) and axon guidance (plexin C1) were altered. These results were reflected by pathway analysis and exemplarily confirmed by PCR and immunohistochemical studies. Supplementing cultured HFs with glutamine or plexin C1 revealed biological relevance and pharmacointerventional potential of these microarray results in altering the HF aging process. Together, we present intriguing data obtained from intra-individual sample comparison that suggest the graying HF to be a valid aging model and a promising target for testing therapeutic interventions.
Sigma-Aldrich
Glutaminase from Escherichia coli, Grade VII, lyophilized powder, 500-1,500 units/mg protein
Sigma-Aldrich
Glutaminase from Escherichia coli, Grade V, lyophilized powder, 50-200 units/mg protein