Merck
  • Efficacy and safety of collagenase clostridium histolyticum injection for Dupuytren contracture: short-term results from 2 open-label studies.
Efficacy and safety of collagenase clostridium histolyticum injection for Dupuytren contracture: short-term results from 2 open-label studies.
The Journal of hand surgery (2012-12-12)
Jörg Witthaut, Graeme Jones, Nebojsa Skrepnik, Harvey Kushner, Anthony Houston, Tommy R Lindau
ABSTRACT
The JOINT I (United States) and JOINT II (Australia and Europe) studies evaluated the efficacy and safety of collagenase clostridium histolyticum (CCH) injection for the treatment of Dupuytren contracture. Both studies used identical open-label protocols. Patients with fixed-flexion contractures of metacarpophalangeal (MCP) (20° to 100°) or proximal interphalangeal (PIP) joints (20° to 80°) could receive up to three 0.58-mg CCH injections per cord (up to 5 total injections per patient). We performed standardized finger extension procedures to disrupt injected cords the next day, with follow-up 1, 2, 6, and 9 months thereafter. The primary end point (clinical success) was reduction in contracture to within 0° to 5° of full extension 30 days after the last injection. Clinical improvement was defined as 50% or more reduction from baseline contracture. Dupuytren cords affecting 879 joints (531 MCP and 348 PIP) in 587 patients were administered CCH injections at 14 U.S. and 20 Australian/European sites, with similar outcomes in both studies. Clinical success was achieved in 497 (57%) of treated joints using 1.2 ± 0.5 (mean ± SD) CCH injections per cord. More MCP than PIP joints achieved clinical success (70% and 37%, respectively) or clinical improvement (89% and 58%, respectively). Less severely contracted joints responded better than those more severely contracted. Mean change in contracture was 55° for MCP joints and 25° for PIP joints. With average contracture reductions of 73% and improvements in range of motion by 30°, most patients (92%) were "very satisfied" (71%) or "quite satisfied" (21%) with treatment. Physicians rated change from baseline as "very much improved" (47%) or "much improved" (35%). The CCH injections were well tolerated, causing no tendon ruptures or systemic reactions. Collagenase clostridium histolyticum was an effective, minimally invasive option for the treatment of Dupuytren contracture of a broad range of severities. Most treated joints (625 of 879) required a single injection. Treatment earlier in the course of disease provided improved outcomes. Therapeutic IV.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, Type V-S, ≥1 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat hepatocytes, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, purified by chromatography, ≥500 CDU/mg solid (CDU = collagen digestion units), lyophilized powder
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, suitable for release of physiologically active rat epididymal adipocytes, Type II-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type L, ≤1.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2 μm filtered solution), suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, non-sterile; 0.2 μm filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, lyophilized powder (from 0.2μm filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type F, ≥2.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, powder, suitable for cell culture, ≥4 FALGPA units/mg solid, high purity, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, 0.2 μm filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, ≥700 CDU/mg solid (CDU = collagen digestion units)
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid