Merck
  • Dehydroepiandrosterone effect on Plasmodium falciparum and its interaction with antimalarial drugs.
Dehydroepiandrosterone effect on Plasmodium falciparum and its interaction with antimalarial drugs.
Experimental parasitology (2012-11-28)
L Zuluaga, S Parra, E Garrido, R López-Muñoz, J D Maya, S Blair
ABSTRACT
Dehydroepiandrosterone (DHEA) inhibits glucose 6-phosphate dehydrogenase (G6PDH) of different species and may potentially decrease intracellular glutathione. Therefore, it can have and enhance anti-parasitic action against Plasmodium spp. We evaluated the antiplasmodial activity and the interaction of DHEA with several antimalarial drugs. The inhibitory effect of DHEA on erythrocytic and G6PDH activity and changes in the content of total and reduced gluthatione Plasmodium falciparum content were also evaluated. DHEA showed antiplasmodial activity in vitro, but the potency was low (IC(50) 118.5 μM). DHEA inhibits G6PDH from healthy erythrocyte and decreases GSH content in both erythrocytes and P. falciparum. DHEA did not synergize or antagonize the antiplasmodial effect of several antimalarial drugs. The most important actions of DHEA were the inhibition of G6PDH activity, and the decrease in both P. falciparum and erythrocyte GSH. Since most of the GSH in Plasmodium spp. infected erythrocytes comes from the parasite itself, it is possible that DHEA analogs could act with higher selectivity, better potency, and might interact synergistically with antimalarials drugs.
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, ≥550 units/mg protein (biuret)