Merck
  • Rosmarinus officinalis polyphenols produce anti-depressant like effect through monoaminergic and cholinergic functions modulation.
Rosmarinus officinalis polyphenols produce anti-depressant like effect through monoaminergic and cholinergic functions modulation.
Behavioural brain research (2012-10-23)
Kazunori Sasaki, Abdelfatteh El Omri, Shinji Kondo, Junkyu Han, Hiroko Isoda
ABSTRACT
Rosmarinus officinalis (R. officinalis), a culinary aromatic and medicinal plant, is very rich in polyphenols and flavonoids with high antioxidant properties. This plant was reported to exert multiple benefits for neuronal system and alleviate mood disorder. In our previous study, we demonstrated that R. officinalis and its active compounds, luteolin (Lut), carnosic acid (CA), and rosmarinic acid (RA), exhibited neurotrophic effects and improved cholinergic functions in PC12 cells in correlation with mitogen-activated protein kinase (MAPK), ERK1/2 signaling pathway. The current study was conducted to evaluate and understand the anti-depressant effect of R. officinalis using tail suspension test (TST) in ICR mice and PC12 cells as in vitro neuronal model. Proteomics analysis of PC12 cells treated with R. officinalis polyphenols (ROP) Lut, CA, and RA revealed a significant upregulation of tyrosine hydroxylase (TH) and pyruvate carboxylase (PC) two major genes involved in dopaminergic, serotonergic and GABAergic pathway regulations. Moreover, ROP were demonstrated to protect neuronal cells against corticosterone-induced toxicity. These results were concordant with decreasing immobility time in TST and regulation of several neurotransmitters (dopamine, norepinephrine, serotonin and acetylcholine) and gene expression in mice brain like TH, PC and MAPK phosphatase (MKP-1). To the best of our knowledge this is the first evidence to contribute to the understanding of molecular mechanism behind the anti-depressant effect of R. officinalis and its major active compounds.
Product Description
Rosmarinic acid, primary reference standard
Pricing and availability is not currently available.
Sigma-Aldrich
Pyruvate Carboxylase from bovine liver, buffered aqueous glycerol solution, 5-25 units/mg protein (BCA)
Pricing and availability is not currently available.
Sigma-Aldrich
Rosmarinic acid, 96%
Pricing and availability is not currently available.
Sigma-Aldrich
Rosmarinic acid, ≥98% (HPLC), from Rosemarinus officinalis L.
Pricing and availability is not currently available.