Merck
  • Predominance of hybrid discrete typing units of Trypanosoma cruzi in domestic Triatoma infestans from the Bolivian Gran Chaco region.
Predominance of hybrid discrete typing units of Trypanosoma cruzi in domestic Triatoma infestans from the Bolivian Gran Chaco region.
Infection, genetics and evolution : journal of molecular epidemiology and evolutionary genetics in infectious diseases (2012-10-11)
Esdenka Perez, Marcelo Monje, Boris Chang, Rosio Buitrago, Rudy Parrado, Christian Barnabé, François Noireau, Simone Frédérique Brenière
ABSTRACT
In the Gran Chaco region the reinfestation by Triatoma infestans remains a major problem for control of Chagas disease. Trypanosoma cruzi the agent of the illness presents a broad genetic intraspecific variability which is poorly documented in the Bolivian Gran Chaco. This work presents the identification of the discrete typing units (DTUs) currently recognized for T. cruzi in T. infestans populations collected before and after residual insecticide spraying in four villages in this region. Before spraying, of 84 samples, the frequencies of the DTUs identified by using the multiplex PCR based on the non transcribed spacer of the mini-exon gene (MMPCR) were 0.21 for TcI, 0.70 for TcII/TcV/TcVI, and 0.17 for TcIII/TcIV and no significant difference was observed after spraying (76 samples). Moreover 13% of the total sample corresponds to T. infestans specimens with mixed infection of DTUs of which three were TcII/TcV/TcVI with TcIII/TcIV. The partial sequences of T. cruzi Gpi gene obtained from 14 PCR products agree the MMPCR DTU identification and allowed to precise the occurrence of TcIII, TcII and hybrid TcV/TcVI stocks which were not discriminated by the MMPCR. Given the high prevalence of hybrid stocks, the authors ask whether the recombination event at the origin of hybrids would have taken place in the Gran Chaco where the putative parents are also present.
Sigma-Aldrich
Phosphoglucose Isomerase from Bacillus stearothermophilus, lyophilized powder, 300-1,000 units/mg protein
Sigma-Aldrich
Phosphoglucose Isomerase from rabbit muscle, Type XI, lyophilized powder, ≥200 units/mg protein
Supelco
Phosphoglucose Isomerase from baker′s yeast (S. cerevisiae), for use with Fructose Assay Kit FA-20
Sigma-Aldrich
Phosphoglucose Isomerase from baker′s yeast (S. cerevisiae), Type III, ammonium sulfate suspension, ≥400 units/mg protein (biuret)