Merck
  • Live diatom silica immobilization of multimeric and redox-active enzymes.
Live diatom silica immobilization of multimeric and redox-active enzymes.
Applied and environmental microbiology (2011-11-08)
V C Sheppard, A Scheffel, N Poulsen, N Kröger
ABSTRACT
Living organisms are adept in forming inorganic materials (biominerals) with unique structures and properties that exceed the capabilities of engineered materials. Biomimetic materials syntheses are being developed that aim at replicating the advantageous properties of biominerals in vitro and endow them with additional functionalities. Recently, proof-of-concept was provided for an alternative approach that allows for the production of biomineral-based functional materials in vivo. In this approach, the cellular machinery for the biosynthesis of nano-/micropatterned SiO₂ (silica) structures in diatoms was genetically engineered to incorporate a monomeric, cofactor-independent ("simple") enzyme, HabB, into diatom silica. In the present work, it is demonstrated that this approach is also applicable for enzymes with "complex" activity requirements, including oligomerization, metal ions, organic redox cofactors, and posttranslational modifications. Functional expression of the enzymes β-glucuronidase, glucose oxidase, galactose oxidase, and horseradish peroxidase in the diatom Thalassiosira pseudonana was accomplished, and 66 to 78% of the expressed enzymes were stably incorporated into the biosilica. The in vivo incorporated enzymes represent approximately 0.1% (wt/wt) of the diatom biosilica and are stabilized against denaturation and proteolytic degradation. Furthermore, it is demonstrated that the gene construct for in vivo immobilization of glucose oxidase can be utilized as the first negative selection marker for diatom genetic engineering.
Sigma-Aldrich
Galactose Oxidase from Dactylium dendroides, ≥30 units/mg solid
Sigma-Aldrich
Galactose Oxidase from Dactylium dendroides, lyophilized powder, ≥3,000 units/g solid