Merck
  • Effect of the conceptus on uterine prostaglandin-F2alpha and prostaglandin-E2 release and synthesis during the periimplantation period in the pig.
Effect of the conceptus on uterine prostaglandin-F2alpha and prostaglandin-E2 release and synthesis during the periimplantation period in the pig.
Reproduction, fertility, and development (2009-06-03)
Marta Wasielak, Katarzyna Kamińska, Marek Bogacki
ABSTRACT
The present study was conducted to evaluate the effect of the conceptus on uterine prostaglandin-F2alpha (PGF2alpha) and prostaglandin-E2 (PGE2) release and the expression of prostaglandin synthase enzymes during the periimplantation period in the pig. A surgically generated model with conceptuses developing in only one of the uterine horns was created. The highest concentration of PGF2alpha and PGE2 was found in the gravid uterine horn, compared with the non-gravid horn and the intact horn of cyclic gilts. Endometrial concentration of both PGs in pregnant gilts was elevated regardless of the conceptus in the uterine horn, whereas only myometrial PGE2 concentration increased during pregnancy. Expression of prostaglandin-E2 synthase (mPGES-1) mRNA in the endometrium was upregulated during the oestrous cycle, while protein expression presented a similar pattern to that of PGE2 concentration in the uterine flushings. Prostaglandin-F2alpha synthase (PGFS) mRNA and protein expression in the endometrium did not differ between pregnancy and oestrous cycle but PGFS mRNA in the myometrium increased during pregnancy both in the gravid and the non-gravid uterine horns. We suggest a local effect of the conceptus on PG release pathways but also a more systemic effect within the whole uterus with regard to PG synthesis and accumulation in uterine tissues.
Sigma-Aldrich
Prostaglandin F Synthase human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)